Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picene (purified by sublimation) (>99.9%) 99.9+%, TCI America™
SDP

Catalog No. P2207100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2207100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2207100MG Supplier TCI America Supplier No. P2207100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 213-46-7
Molecular Formula C22H14
Molecular Weight (g/mol) 278.354
MDL Number MFCD00215857
InChI Key GBROPGWFBFCKAG-UHFFFAOYSA-N
Synonym 3,4-benzchrysene, pycene, benzo a chrysene, 1,2:7,8-dibenzophenanthrene, beta,beta-binaphthyleneethene, dibenzo a,i phenanthrene, 1,2,7,8-dibenzphenanthrene, unii-f70r8zbr7t, f70r8zbr7t
PubChem CID 9162
ChEBI CHEBI:33090
IUPAC Name picene
SMILES C1=CC=C2C(=C1)C=CC3=C2C=CC4=C3C=CC5=CC=CC=C54

Specifications

Melting Point 366°C
Color Yellow
Boiling Point 525°C
Quantity 100 mg
Formula Weight 278.35
Percent Purity ≥99.9% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Picene (purified by sublimation) (>99.9%)
RTECSNumber TJ3700000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.