accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2131-41-1

IUPAC Name: 1,4,5-trimethylnaphthalene
Synonyms: 1,4,5-trimethylnaphthalene
Molecular Weight (g/mol): 170.26
Molecular Formula: C13H14
InChi Key: FSAWRQYDMHSDRN-UHFFFAOYSA-N
SMILES: CC1=CC=CC2=C(C)C=CC(C)=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results