Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4,5-Trimethylnaphthalene 96.0+%, TCI America™
SDP

Catalog No. T1711100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1711100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1711100MG Supplier TCI America Supplier No. T1711100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2131-41-1
Molecular Formula C13H14
Molecular Weight (g/mol) 170.255
MDL Number MFCD00216209
InChI Key FSAWRQYDMHSDRN-UHFFFAOYSA-N
PubChem CID 16478
IUPAC Name 1,4,5-trimethylnaphthalene
SMILES CC1=C2C=CC=C(C2=C(C=C1)C)C

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 145°C
Quantity 100 mg
Formula Weight 170.26
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,5-Trimethylnaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.