accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHH2NH2O

CAS RN 213178-94-0

IUPAC Name: 2-amino-3-cyclohexylpropanoic acid hydrate
Synonyms: cyclohexylalanine hydrate
Molecular Weight (g/mol): 189.26
Molecular Formula: C9H19NO3
InChi Key: HDJQIWAIDCEDEF-UHFFFAOYNA-N
SMILES: O.NC(CC1CCCCC1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results