accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 213203-65-7

FBrFFF

CAS RN 213203-65-7

IUPAC Name: 4-(bromomethyl)-2-fluoro-1-(trifluoromethyl)benzene
Synonyms: 4-(bromomethyl)-2-fluoro-1-(trifluoromethyl)benzene
Molecular Weight (g/mol): 257.03
Molecular Formula: C8H5BrF4
InChi Key: MOVSRIIBGRAUAF-UHFFFAOYSA-N
SMILES: FC1=CC(CBr)=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results