accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21351-79-1

IUPAC Name: caesium(1+) hydroxide
Synonyms: cesium hydroxide|CsOH|cesium hydrate
Molecular Weight (g/mol): 149.91
Molecular Formula: CsHO
InChi Key: HUCVOHYBFXVBRW-UHFFFAOYSA-M
SMILES: [OH-].[Cs+]
Percent Purity 
  • (2)
  • (2)
  • (4)
  • (3)
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (6)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)