Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cesium hydroxide solution

Catalog No. 23204150G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
200 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-204-150G 50 g
23-204-1200G 200 g
23-204-110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-204-150G Supplier Sigma Aldrich Supplier No. 23204150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cesium hydroxide solution is used to make porous cesium titanosilicates, potential ion-exchange materials for cleanup of radioactive wastes.CsOH impregnated in FAU zeolites enhanced their basic properties without affecting the FAU framework.It can effectively catalyse the condensation of methyl propionate with formaldehyde to yield methyl methacrylate.

Specifications

CAS 21351-79-1
Density 1.72 g/mL (at 25°C)
Molecular Formula HCsO
Linear Formula CsOH
Quantity 50 g
MDL Number MFCD00010964
Synonym Caesium hydroxide
Molecular Weight (g/mol) 149.91
Percent Purity 99.9% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.