accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOClClOHOClCl

CAS RN 2136-79-0

IUPAC Name: tetrachlorobenzene-1,4-dicarboxylic acid
Synonyms: tetrachlorobenzene-1,4-dicarboxylic acid
Molecular Weight (g/mol): 303.9
Molecular Formula: C8H2Cl4O4
InChi Key: KZCBXHSWMMIEQU-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C(Cl)=C(C(O)=O)C(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results