CAS RN 21366-63-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1000 μg/mL Cannabicyclol (CBL) Standard in P&T Methanol, Restek
Ideal for creating multipoint (5-point min suggested) calibration curves for LC analysis