Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1000 μg/mL Cannabicyclol (CBL) Standard in P&T Methanol, Restek
SDP

Catalog No. 06611897 Shop All Restek Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-611-897 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Must ship on ice
Catalog No. 06-611-897 Supplier Restek Corporation Supplier No. 34130
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Must ship on ice

Ideal for creating multipoint (5-point min suggested) calibration curves for LC analysis

  • Verified composition and stability
  • For accurate quantification in cannnabis testing and chemovar ID
  • Prepared stock product eliminates need for in-house standards prep
  • Pair with Restek's cannabinoid standards for a more comprehensive analysis of cannabis products by LC
  • Excluded from U.S. DEA Controlled Substances Act (CSA) regulatory controls
    • No customer permits or licensing required to purchase within U.S.

Specifications

CAS 21366-63-2
Shipping Condition On ice
Concentration 1000 μg/mL
Certifications/Compliance Certified reference material
Type Cannabinoids Standard
Solvent or Matrix Methanol-P and T
Industry Type Cannabis Testing
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.