accessibility menu, dialog, popup

UserName

Viewing profile as user:
IH3CNCH3H3CCH3

CAS RN 2138-24-1

IUPAC Name: tetrahexylazanium iodide
Synonyms: tetrahexylazanium iodide
Molecular Weight (g/mol): 481.59
Molecular Formula: C24H52IN
InChi Key: VRKHAMWCGMJAMI-UHFFFAOYSA-M
SMILES: [I-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)