CAS RN 21382-98-9
IUPAC Name:
4-(methylsulfanyl)benzonitrile
Synonyms:
4-(methylsulfanyl)benzonitrile
Molecular Weight (g/mol):
149.21
Molecular Formula:
C8H7NS
InChi Key:
RPVGEEHGKIFQFO-UHFFFAOYSA-N
SMILES:
CSC1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
Sigma Aldrich 4-(Methylthio)benzonitrile
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.