accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OHOOONNONNH

CAS RN 2140-11-6

IUPAC Name: 9-[6-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl]-6,9-dihydro-3H-purin-6-one
Synonyms: 9-[6-(hydroxymethyl)-2,2-dimethyl-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]-3H-purin-6-one
Molecular Weight (g/mol): 308.29
Molecular Formula: C13H16N4O5
InChi Key: LIEKLUBCIPVWQD-UHFFFAOYNA-N
SMILES: CC1(C)OC2C(CO)OC(C2O1)N1C=NC2=C1NC=NC2=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)