Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3'-O-Isopropylideneinosine 98.0+%, TCI America™
SDP

Catalog No. I07045G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I07045G Supplier TCI America Supplier No. I07045G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2140-11-6
Molecular Formula C13H16N4O5
Molecular Weight (g/mol) 308.29
MDL Number MFCD00038002
InChI Key LIEKLUBCIPVWQD-UHFFFAOYNA-N
Synonym 9-(2,3-O-Isopropylidene-beta-D-ribofuranosyl)hypoxanthine
PubChem CID 129317592
IUPAC Name 9-[(3aS,6aR)-6-(hydroxymethyl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]-3H-purin-6-one
SMILES CC1(OC2C(OC(C2O1)N3C=NC4=C3NC=NC4=O)CO)C

Specifications

Quantity 5 g
Formula Weight 308.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3′-O-Isopropylideneinosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.