accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(R)N(R)O(S)(R)OOHNH2HOHOCH3

CAS RN 2140-61-6

IUPAC Name: 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2-dihydropyrimidin-2-one
Synonyms: 5-methylcytidine
Molecular Weight (g/mol): 257.25
Molecular Formula: C10H15N3O5
InChi Key: ZAYHVCMSTBRABG-JXOAFFINSA-N
SMILES: CC1=CN([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N=C1N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results