accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2142-01-0

IUPAC Name: 2-benzyl-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: N-benzylphthalimide
Molecular Weight (g/mol): 237.26
Molecular Formula: C15H11NO2
InChi Key: WITXFYCLPDFRNM-UHFFFAOYSA-N
SMILES: O=C1N(CC2=CC=CC=C2)C(=O)C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results