Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylphthalimide 98.0+%, TCI America™
SDP

Catalog No. B199725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B199725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B199725G Supplier TCI America Supplier No. B199725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2142-01-0
Molecular Formula C15H11NO2
Molecular Weight (g/mol) 237.26
MDL Number MFCD00023077
InChI Key WITXFYCLPDFRNM-UHFFFAOYSA-N
PubChem CID 75059
ChEBI CHEBI:34866
IUPAC Name 2-benzyl-2,3-dihydro-1H-isoindole-1,3-dione
SMILES O=C1N(CC2=CC=CC=C2)C(=O)C2=CC=CC=C12

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 25 g
Formula Weight 237.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzylphthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.