accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHONNNOOO

CAS RN 214467-60-4

IUPAC Name: N-[2-azido-4,5-bis(benzyloxy)-6-[(benzyloxy)methyl]oxan-3-yl]acetamide
Synonyms: N-[2-azido-4,5-bis(benzyloxy)-6-[(benzyloxy)methyl]oxan-3-yl]acetamide
Molecular Weight (g/mol): 516.6
Molecular Formula: C29H32N4O5
InChi Key: XOOBJDMHDWWHLZ-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1)N=[N+]=[N-]
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)