accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHOOONHNH(R)H2C

CAS RN 214750-74-0

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-{[(prop-2-en-1-yloxy)carbonyl]amino}pentanoic acid
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-{[(prop-2-en-1-yloxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 438.48
Molecular Formula: C24H26N2O6
InChi Key: RXLIOYNXBHZZBI-OAQYLSRUSA-N
SMILES: OC(=O)[C@@H](CCCNC(=O)OCC=C)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)