accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 214750-76-2

IUPAC Name: (2S)-3-cyclopropyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2S)-3-cyclopropyl-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 351.4
Molecular Formula: C21H21NO4
InChi Key: DRGUEWQZLABTFG-IBGZPJMESA-N
SMILES: OC(=O)[C@H](CC1CC1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results