accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 215-58-7

IUPAC Name: pentacyclo[12.8.0.0²,⁷.0⁸,¹³.0¹⁶,²¹]docosa-1(22),2,4,6,8,10,12,14,16,18,20-undecaene
Synonyms: 1,2,3,4-dibenzanthracene
Molecular Weight (g/mol): 278.35
Molecular Formula: C22H14
InChi Key: RAASUWZPTOJQAY-UHFFFAOYSA-N
SMILES: C1=CC=C2C=C3C4=CC=CC=C4C4=CC=CC=C4C3=CC2=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)