Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Dibenzanthracene, 97%, Thermo Scientific™

Catalog No. AC112505000
Change view
Click to view available options
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC112505000 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC112505000 Supplier Thermo Scientific Chemicals Supplier No. 112505000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 215-58-7
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C22H14
Quantity 500mg
MDL Number MFCD00003603
Beilstein 05, II, 668
Synonym benzo f tetraphene, benzo b triphenylene, dibenz a,c anthracene, dibenzo a,c anthracene, benzotriphenylene, 1,2:3,4-dibenzanthracene, 1,2,3,4-dibenzanthracene
InChI Key RAASUWZPTOJQAY-UHFFFAOYSA-N
SMILES C1=CC=C2C=C3C4=CC=CC=C4C4=CC=CC=C4C3=CC2=C1
IUPAC Name pentacyclo[12.8.0.0²,⁷.0⁸,¹³.0¹⁶,²¹]docosa-1(22),2,4,6,8,10,12,14,16,18,20-undecaene
Molecular Weight (g/mol) 278.35
PubChem CID 9164
ChEBI CHEBI:82437
Formula Weight 278.34
Percent Purity ≥96.0%
Chemical Name or Material 1,2,3,4-Dibenzanthracene, 97%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Suspected of causing genetic defects.
Suspected of causing cancer.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-920-8
RTECSNumber DM1925000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.