accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3OH3C

CAS RN 2150-37-0

IUPAC Name: methyl 3,5-dimethoxybenzoate
Synonyms: methyl 3,5-dimethoxybenzoate
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: YXUIOVUOFQKWDM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OC)=CC(OC)=C1
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)