accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21535-47-7

IUPAC Name: hydrogen 5-methyl-2,5-diazatetracyclo[13.4.0.0²,⁷.0⁸,¹³]nonadeca-1(19),8,10,12,15,17-hexaene chloride
Synonyms: mianserin hydrochloride mianserine hydrochloride
Molecular Weight (g/mol): 300.83
Molecular Formula: C18H21ClN2
InChi Key: YNPFMWCWRVTGKJ-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CN1CCN2C(C1)C1=CC=CC=C1CC1=CC=CC=C21
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)