accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2155-74-0

IUPAC Name: antimony(3+) tris(butan-1-olate)
Synonyms: antimony(3+) tris(butan-1-olate)
Molecular Weight (g/mol): 341.11
Molecular Formula: C12H27O3Sb
InChi Key: YGBFTDQFAKDXBZ-UHFFFAOYSA-N
SMILES: [Sb+3].CCCC[O-].CCCC[O-].CCCC[O-]
Boiling Point 
  • (3)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)