accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21553-46-8

IUPAC Name: 4-methoxy-3,5-dimethylbenzoate
Synonyms: 4-methoxy-3,5-dimethylbenzoate
Molecular Weight (g/mol): 179.2
Molecular Formula: C10H11O3
InChi Key: WXVQURJGDUNJCS-UHFFFAOYSA-M
SMILES: COC1=C(C)C=C(C=C1C)C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)