accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 215589-24-5

NOF

CAS RN 215589-24-5

IUPAC Name: 4-(4-fluorophenoxy)benzonitrile
Synonyms: 4-(4-fluorophenoxy)benzonitrile
Molecular Weight (g/mol): 213.21
Molecular Formula: C13H8FNO
InChi Key: NXVPHQNXBDDVCT-UHFFFAOYSA-N
SMILES: FC1=CC=C(OC2=CC=C(C=C2)C#N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results