Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Fluorophenoxy)benzonitrile 98.0+%, TCI America™
SDP

Catalog No. F05325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05325G 5 g
F05321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05325G Supplier TCI America Supplier No. F05325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 215589-24-5
Molecular Formula C13H8FNO
Molecular Weight (g/mol) 213.211
InChI Key NXVPHQNXBDDVCT-UHFFFAOYSA-N
Synonym 4-4-fluorophenoxy benzonitrile, 4-4-fluorophenoxy benzenecarbonitrile, acmc-1chxs, 4-cyano-4'-fluorodiphenyl ether, benzonitrile,4-4-fluorophenoxy
PubChem CID 1490284
IUPAC Name 4-(4-fluorophenoxy)benzonitrile
SMILES C1=CC(=CC=C1C#N)OC2=CC=C(C=C2)F

Specifications

Melting Point 69°C
Color White
Quantity 5 g
Formula Weight 213.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Fluorophenoxy)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.