accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHHNFFFNH2

CAS RN 215655-42-8

IUPAC Name: tert-butyl N-(2-{[2-amino-4-(trifluoromethyl)phenyl]amino}ethyl)carbamate
Synonyms: tert-butyl N-(2-{[2-amino-4-(trifluoromethyl)phenyl]amino}ethyl)carbamate
Molecular Weight (g/mol): 319.33
Molecular Formula: C14H20F3N3O2
InChi Key: LBPALYCQELHDIC-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCNC1=C(N)C=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results