Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl N-{2-[2-amino-4-(trifluoromethyl)anilino]ethyl}carbamate, 95%, Thermo Scientific™

Catalog No. AW00453DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AW00453DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AW00453DA Supplier Thermo Scientific Chemicals Supplier No. AW00453DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 215655-42-8
Molecular Formula C14H20F3N3O2
Molecular Weight (g/mol) 319.328
MDL Number MFCD00728760
InChI Key LBPALYCQELHDIC-UHFFFAOYSA-N
Synonym tert-butyl n-2-2-amino-4-trifluoromethyl anilino ethyl carbamate, tert-butyl n-2-2-amino-4-trifluoromethyl phenyl amino ethyl carbamate, tert-butyl 2-2-amino-4-trifluoromethyl phenyl amino ethylcarbamate, tert-butyl-n-2-2-amino-4-trifluoromethyl anilino ethyl carbamate, n-2-2-amino-4-trifluoromethyl phenyl amino ethyl tert-butoxy carboxamide, maybridge1_000225, tert-butyl 2-2-amino-4-trifluoromethyl anilino ethyl carbamate, t-butyl n-2-2-amino-4-trifluoromethyl anilino ethylcarbamate
PubChem CID 2773437
IUPAC Name tert-butyl N-[2-[2-amino-4-(trifluoromethyl)anilino]ethyl]carbamate
SMILES CC(C)(C)OC(=O)NCCNC1=C(C=C(C=C1)C(F)(F)F)N

Specifications

CAS Min % 94.5
CAS Max % 95.0
Assay Percent Range 95%
Linear Formula CF3C6H3(NH2)NH(CH2)2NHC(O)OC(CH3)3
Quantity 1 g
Formula Weight 319.33
Percent Purity 95%
Chemical Name or Material tert-Butyl N-{2-[2-amino-4-(trifluoromethyl)anilino]ethyl}carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.