CAS RN 215788-65-1
IUPAC Name:
Dichloro[1,1'-bis(diisopropylphosphino)ferrocene]palladium(II)
Synonyms:
—
Molecular Weight (g/mol):
595.64
Molecular Formula:
C22H36Cl2FeP2Pd
InChi Key:
—
SMILES:
[Fe].Cl[Pd]Cl.CC(C)P(C(C)C)c1cccc1.CC(C)P(C(C)C)c1cccc1
1
–
1
of
1
results
1
Sigma Aldrich (1R,2R)-(+)-N,N'-Dimethyl-1,2-Bis[3-(Trifluoromethyl)Phenyl]Ethanediamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.