accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2158-08-9

IUPAC Name: octadecylurea
Synonyms: octadecylurea
Molecular Weight (g/mol): 312.54
Molecular Formula: C19H40N2O
InChi Key: GJNDMSSZEBNLPU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCNC(N)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results