Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Octadecylurea 97.0+%, TCI America™
SDP

Catalog No. O020910G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O020910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O020910G Supplier TCI America Supplier No. O020910G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2158-08-9
Molecular Formula C19H40N2O
Molecular Weight (g/mol) 312.54
MDL Number MFCD00043623
InChI Key GJNDMSSZEBNLPU-UHFFFAOYSA-N
PubChem CID 75090
IUPAC Name octadecylurea
SMILES CCCCCCCCCCCCCCCCCCNC(=O)N

Specifications

Melting Point 110°C
Color White
Quantity 10 g
Formula Weight 312.54
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Octadecylurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.