accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CBOHHO

CAS RN 216019-28-2

IUPAC Name: [3-(propan-2-yl)phenyl]boronic acid
Synonyms: 3-isopropylphenylboronic acid
Molecular Weight (g/mol): 164.01
Molecular Formula: C9H13BO2
InChi Key: QSWLFBMVIGQONC-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(=C1)B(O)O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)