Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Isopropylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. I08875G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0887-5G 5 g
I08871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0887-5G Supplier TCI America Supplier No. I08875G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 216019-28-2
Color White
Molecular Formula C9H13BO2
Quantity 5 g
MDL Number MFCD01074634
Synonym 3-isopropylphenylboronic acid, 3-isopropylbenzeneboronic acid, 3-cumylboronic acid, 3-isopropylphenyl boronic acid, 3-isopropylphebylboronic acid
InChI Key QSWLFBMVIGQONC-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)C(C)C)(O)O
IUPAC Name (3-propan-2-ylphenyl)boronic acid
Molecular Weight (g/mol) 164.011
PubChem CID 2773478
Formula Weight 164.01
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.