accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONOOOOH

CAS RN 21606-04-2

IUPAC Name: 2-(methoxycarbonyl)-6-nitrobenzoic acid
Synonyms: 2-(methoxycarbonyl)-6-nitrobenzoic acid
Molecular Weight (g/mol): 225.16
Molecular Formula: C9H7NO6
InChi Key: DJMQLZPEBHSABD-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C(O)=O)C(=CC=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results