CAS RN 216163-53-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PD 174265
A potent, cell-permeable, reversible, ATP-competitive and selective inhibitor of EGF receptor (EGFR) tyrosine kinase activity (IC50 = 450 pM).