Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PD 174265

Catalog No. 5130401MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-304-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-304-01MG Supplier MilliporeSigma™ Supplier No. 5130401MG
Add to Cart
Add to Cart

A potent, cell-permeable, reversible, ATP-competitive and selective inhibitor of EGF receptor (EGFR) tyrosine kinase activity (IC50 = 450 pM).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 97% by HPLC
Target Proteins EGFR tyrosine kinase activity
CAS 216163-53-0
Molecular Formula C17H15BrN4O
Formula Weight 371.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.