accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2162-98-3

IUPAC Name: 1,10-dichlorodecane
Synonyms: decane, 1,10-dichloro-
Molecular Weight (g/mol): 211.17
Molecular Formula: C10H20Cl2
InChi Key: RBBNTRDPSVZESY-UHFFFAOYSA-N
SMILES: ClCCCCCCCCCCCl
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)