Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Dichlorodecane, 98%

Catalog No. AC255930250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC255930250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC255930250 Supplier Thermo Scientific Chemicals Supplier No. 255930250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2162-98-3 | C10H20Cl2 | 211.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2162-98-3
Molecular Formula C10H20Cl2
Molecular Weight (g/mol) 211.17
MDL Number MFCD00000957
InChI Key RBBNTRDPSVZESY-UHFFFAOYSA-N
Synonym decane, 1,10-dichloro, decamethylene dichloride, decane,10-dichloro, 1, 10-dichlorodecane, acmc-1cbxv, ksc202q6p, 1,10-dichlorodecane, 1,10-decamethylene dichloride
PubChem CID 75101
IUPAC Name 1,10-dichlorodecane
SMILES ClCCCCCCCCCCCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 15.6°C
Color Colorless
Density 0.9990g/mL
Boiling Point 275.0°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4600 to 1.4620
Linear Formula Cl(CH2)10Cl
Quantity 25 g
Beilstein 01, 522
Specific Gravity 0.999
Solubility Information Solubility in water: 0.77g/L (20 c)
Viscosity 4.72 mPa.s (20°C)
Formula Weight 211.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 10-Dichlorodecane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 218-489-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.