accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNONNHH3CCH3HN

CAS RN 2163-68-0

IUPAC Name: 4-(ethylamino)-6-[(propan-2-yl)amino]-2,5-dihydro-1,3,5-triazin-2-one
Synonyms: 4-(ethylamino)-6-(isopropylamino)-5H-1,3,5-triazin-2-one
Molecular Weight (g/mol): 197.24
Molecular Formula: C8H15N5O
InChi Key: NFMIMWNQWAWNDW-UHFFFAOYSA-N
SMILES: CCNC1=NC(=O)N=C(NC(C)C)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results