accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBOHBrF

CAS RN 216393-64-5

IUPAC Name: (4-bromo-2-fluorophenyl)boronic acid
Synonyms: 4-bromo-2-fluorophenylboronic acid
Molecular Weight (g/mol): 218.82
Molecular Formula: C6H5BBrFO2
InChi Key: PKZHIIVZRPUZSU-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(Br)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results