Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-fluorobenzeneboronic acid, 98+%, Thermo Scientific™

Catalog No. AAL1746806
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1746806 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1746806 Supplier Thermo Scientific Chemicals Supplier No. L1746806

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is a pharmaceutical intermediate. Arylboronic acids undergo cross-coupling with arenediazonium tetrafluoroborates in dioxane or methanol, catalyzed by palladium acetate in the absence of both added base and phosphine ligand.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 216393-64-5
Molecular Formula C6H5BBrFO2
Quantity 5g
MDL Number MFCD01318110
Synonym 4-bromo-2-fluorobenzeneboronic acid, 4-bromo-2-fluorophenyl boronic acid, 1-borono-4-bromo-2-fluorobenzene, pubchem7778, acmc-209fms, 4-bromo-2-fluorobenzeneboronicacid, 4-bromo-2-fluoro-phenylboronic acid, 4-bromo-2-fluoro-phenyl boronic acid, boronic acid, 4-bromo-2-fluorophenyl
Solubility Information Insoluble in water.
InChI Key PKZHIIVZRPUZSU-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(Br)C=C1F
IUPAC Name (4-bromo-2-fluorophenyl)boronic acid
Molecular Weight (g/mol) 218.82
PubChem CID 2773333
Formula Weight 218.82
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-2-fluorobenzeneboronic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.