accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 216431-85-5

NNFFF

CAS RN 216431-85-5

IUPAC Name: 6-(trifluoromethyl)pyridine-3-carbonitrile
Synonyms: 6-(trifluoromethyl)pyridine-3-carbonitrile
Molecular Weight (g/mol): 172.11
Molecular Formula: C7H3F3N2
InChi Key: ZBLULFJVRGVQAW-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC=C(C=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results