Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Cyano-2-(trifluoromethyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. C26981G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C26981G Supplier TCI America Supplier No. C26981G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 216431-85-5 | C7H3F3N2 | 172.11 g/mol

Chemical Identifiers

CAS 216431-85-5
Molecular Formula C7H3F3N2
Molecular Weight (g/mol) 172.11
MDL Number MFCD03001347
InChI Key ZBLULFJVRGVQAW-UHFFFAOYSA-N
Synonym 6-trifluoromethyl nicotinonitrile, 5-cyano-2-trifluoromethyl pyridine, 6-trifluoromethyl pyridine-3-carbonitrile, 6-trifluoromethyl-nicotinonitrile, 3-cyano-6-trifluoromethylpyridine, 3-pyridinecarbonitrile, 6-trifluoromethyl, 2-trifluoromethyl-5-cyanopyridine, pubchem14525, acmc-209fmx, ksc495e8b
PubChem CID 2777554
IUPAC Name 6-(trifluoromethyl)pyridine-3-carbonitrile
SMILES C1=CC(=NC=C1C#N)C(F)(F)F

Specifications

Melting Point 38°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 172.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Cyano-2-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.