accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNNH3CCH3SOOO

CAS RN 216667-49-1

IUPAC Name: 3-[dimethyl({3-[(4-octylphenyl)formamido]propyl})azaniumyl]propane-1-sulfonate
Synonyms: 3-[dimethyl({3-[(4-octylphenyl)formamido]propyl})ammonio]propane-1-sulfonate
Molecular Weight (g/mol): 440.64
Molecular Formula: C23H40N2O4S
InChi Key: AQVJFUZGGMXMED-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=C(C=C1)C(=O)NCCC[N+](C)(C)CCCS([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results