Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ASB-C8Ø, Calbiochem™,

Catalog No. 1827305GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-273-05GM 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-273-05GM Supplier MilliporeSigma™ Supplier No. 1827305GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 216667-49-1
Molecular Formula C23H40N2O4S
Molecular Weight (g/mol) 440.643
InChI Key AQVJFUZGGMXMED-UHFFFAOYSA-N
Synonym C8Ø, 4-n-Octylbenzoylamido-propyl-dimethylammonio Sulfobetaine
PubChem CID 3281612
IUPAC Name 3-[dimethyl-[3-[(4-octylbenzoyl)amino]propyl]azaniumyl]propane-1-sulfonate
SMILES CCCCCCCCC1=CC=C(C=C1)C(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-]

Specifications

Color White
Quantity 5 g
Packaging Glass Bottle
Percent Purity ≥95% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.