CAS RN 2167-51-3
IUPAC Name:
4-(2-{4-[2-(4-hydroxyphenyl)propan-2-yl]phenyl}propan-2-yl)phenol
Synonyms:
4-(2-{4-[2-(4-hydroxyphenyl)propan-2-yl]phenyl}propan-2-yl)phenol
Molecular Weight (g/mol):
346.47
Molecular Formula:
C24H26O2
InChi Key:
GIXXQTYGFOHYPT-UHFFFAOYSA-N
SMILES:
CC(C)(C1=CC=C(O)C=C1)C1=CC=C(C=C1)C(C)(C)C1=CC=C(O)C=C1
1
–
1
of
1
results
1
alpha,alpha'-Bis(4-hydroxyphenyl)-1,4-diisopropylbenzene 98.0+%, TCI America™
CAS: 2167-51-3 Molecular Formula: C24H26O2 Molecular Weight (g/mol): 346.47 MDL Number: MFCD00191315 InChI Key: GIXXQTYGFOHYPT-UHFFFAOYSA-N PubChem CID: 630355 IUPAC Name: 4-[2-[4-[2-(4-hydroxyphenyl)propan-2-yl]phenyl]propan-2-yl]phenol SMILES: CC(C)(C1=CC=C(C=C1)C(C)(C)C2=CC=C(C=C2)O)C3=CC=C(C=C3)O