accessibility menu, dialog, popup

UserName

Viewing profile as user:
FBrBr

CAS RN 216755-57-6

IUPAC Name: 1-bromo-3-(bromomethyl)-5-fluorobenzene
Synonyms: 1-bromo-3-(bromomethyl)-5-fluorobenzene
Molecular Weight (g/mol): 267.92
Molecular Formula: C7H5Br2F
InChi Key: DAUWIPUGOIFZNF-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC(CBr)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results