Learn More
3-Bromo-5-fluorobenzyl bromide, 95%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
3-Bromo-5-fluorobenzyl bromide is employed as a intermediate for pharmaceutical and chemical research.
Solubility
Slightly soluble in water (2.9 g/L) (25°C).
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Specifications
| Quantity | 1 g |
| UN Number | UN3261 |
| Solubility Information | Slightly soluble in water (2.9g/L) (25°C). |
| Formula Weight | 267.92 |
| Percent Purity | 95% |
| Chemical Name or Material | 3-Bromo-5-fluorobenzyl bromide |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Keep cold |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.